Skip to Content
Merck
All Photos(1)

Key Documents

C6734

Sigma-Aldrich

CAY10397

solid, >98%

Synonym(s):

5-[[4-(Ethoxycarbonyl)phenyl]azo]-2-hydroxybenzeneacetic acid, CK47A

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$225.00

$225.00


Estimated to ship onMay 17, 2025



Select a Size

Change View
1 MG
$225.00

About This Item

Empirical Formula (Hill Notation):
C17H16N2O5
CAS Number:
Molecular Weight:
328.32
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$225.00


Estimated to ship onMay 17, 2025


Product Name

CAY10397, >98%, solid

Quality Level

Assay

>98%

form

solid

shipped in

wet ice

storage temp.

−20°C

SMILES string

CCOC(=O)c1ccc(cc1)\N=N\c2ccc(O)c(CC(O)=O)c2

InChI

1S/C17H16N2O5/c1-2-24-17(23)11-3-5-13(6-4-11)18-19-14-7-8-15(20)12(9-14)10-16(21)22/h3-9,20H,2,10H2,1H3,(H,21,22)/b19-18+

InChI key

YYBSDOZYJSGLTH-VHEBQXMUSA-N

Biochem/physiol Actions

Potent and selective inhibitor of 15-hydroxyprostaglandin dehydrogenase (15-hydroxy-PGDH). Acts to prolong the lifetime and activity of endogenously produced prostaglandins both in vitro and in vivo.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christian Hunnicke Petersen et al.
BMC cancer, 19(1), 643-643 (2019-06-30)
Investigations of colorectal carcinogenesis have mainly focused on examining neoplastic tissue. With our aim of identifying potentially cancer-predisposing molecular compositions, we chose a different approach by examining endoscopically normal appearing colonic mucosa of patients with and without colorectal neoplasia (CRN).
Shaneice K Nettleford et al.
Frontiers in nutrition, 7, 96-96 (2020-08-11)
Enteropathogenic Escherichia coli (EPEC) leads to adverse colonic inflammation associated with poor resolution of inflammation and loss of epithelial integrity. Micronutrient trace element selenium (Se) is incorporated into selenoproteins as the 21st amino acid, selenocysteine (Sec). Previous studies have shown
Liuping Wei et al.
Cellular signalling, 26(7), 1476-1488 (2014-03-25)
We have established that 15-hydroxyeicosatetraenoic acid is an important factor in regulation of pulmonary vascular remodeling (PVR) associated with hypoxia-induced pulmonary hypertension (PH), which is further metabolized by 15-hydroxyprostaglandin dehydrogenase (15-PGDH) to form 15-ketoeicosatetraenoic acid (15-KETE). However, the role of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service