Skip to Content
Merck
All Photos(1)

Key Documents

C6613

Sigma-Aldrich

Anti-Cdc7 Kinase antibody, Mouse monoclonal

clone DCS-341, purified from hybridoma cell culture

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

DCS-341, monoclonal

form

buffered aqueous solution

species reactivity

human

concentration

~2 mg/mL

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
indirect ELISA: suitable
microarray: suitable
western blot: 0.4 μg/mL using total cell extract of A431 cells

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CDC7(8317)

General description

Anti-Cdc7 Kinase antibody, Mouse monoclonal (mouse IgG1 isotype) is derived from the DCS-341 hybridoma produced by the fusion of mouse myeloma cells (NS2) and splenocytes from BALB/c mice immunized with full length human Cdc7 kinase. Cell division cycle 7-related protein kinase(CDC7) is an enzyme encoded by the CDC7 gene in humans. It is highly expressed in diffuse large B cell lymphoma (DLBCL). Human Cdc7 kinase consists of 574 amino acids with a molecular weight of 55 kDa. CDC7 shares nearly 45% identity with yeast Cdc7 kinase in the kinase domain.

Immunogen

full-length human Cdc7 kinase.

Application

Anti-Cdc7 Kinase antibody, Mouse monoclonal has been used in:
  • enzyme-linked immunosorbent assay (ELISA)
  • immunoblotting
  • immunoprecipitation
  • immunocytochemistry.

Biochem/physiol Actions

Cell division cycle 7-related protein kinase (CDC7) in addition to its role in chromosome structure maintenance and repair, is also involved in meiosis and checkpoint responses.
Cell division cycle 7-related protein kinase(CDC7) is a serine-threonine essential kinase required for the initiation of DNA replication and is involved in regulation of the cell cycle. It is high expressed in diffuse large B cell lymphoma (DLBCL). Loss of Cdc7 kinase expression results in retarded growth at both cellular and whole body levels, with especially profound impairment of germ cell development. It inhibits genotoxin-induced apoptosis and increases survival in cancer cells. It may play an important role in the development and progression of colorectal cancer. Depletion of Cdc7 induces cancer cell death regardless of its p53 status. It can also interact with and phosphorylate Claspin.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael D Rainey et al.
Cell cycle (Georgetown, Tex.), 12(10), 1560-1568 (2013-04-20)
Claspin is a critical mediator protein in the DNA replication checkpoint, responsible for ATR-dependent activation of the effector kinase Chk1. Cdc7, an essential kinase required for the initiation of DNA replication, can also interact with and phosphorylate Claspin. In this
Hai-Jun Chen et al.
World journal of gastroenterology, 19(20), 3130-3133 (2013-05-30)
To detect the expression of huCdc7 in colorectal cancer. The mRNA and protein expression of huCdc7 in 39 colorectal cancer tissue specimens and matched tumor-adjacent normal colorectal tissue specimens was detected by reverse transcription-polymerase chain reaction and immunohistochemistry, respectively. The
Increased Activity of the S Phase Kinase Cdc7 Is Associated with Poor Outcome in Diffuse Large B Cell Lymphoma (DLBCL).
Krawczyk J, et al.
Blood (2009)
Jung Min Kim et al.
Mutation research, 532(1-2), 29-40 (2003-12-04)
Cdc7 kinase plays an essential role in firing of replication origins by phosphorylating components of the replication complexes. Cdc7 kinase has also been implicated in S phase checkpoint signaling downstream of the ATR and Chk1 kinases. Inactivation of Cdc7 in
Human and Xenopus cDNAs encoding budding yeast Cdc7-related kinases: in vitro phosphorylation of MCM subunits by a putative human homologue of Cdc7
Sato N, et al.
The Embo Journal, 16(14), 4340-4351 (1997)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service