Skip to Content
Merck
All Photos(1)

Key Documents

C6524

Sigma-Aldrich

Cholesteryl hexanoate

Synonym(s):

3β-Hydroxy-5-cholestene 3-hexanoate, 5-Cholesten-3β-ol 3-hexanoate, Cholesteryl caproate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H56O2
CAS Number:
Molecular Weight:
484.80
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:

storage temp.

−20°C

SMILES string

CCCCCC(=O)O[C@H]1CC[C@]2(C)[C@H]3CC[C@]4(C)[C@H](CC[C@H]4[C@@H]3CC=C2C1)[C@H](C)CCCC(C)C

InChI

1S/C33H56O2/c1-7-8-9-13-31(34)35-26-18-20-32(5)25(22-26)14-15-27-29-17-16-28(24(4)12-10-11-23(2)3)33(29,6)21-19-30(27)32/h14,23-24,26-30H,7-13,15-22H2,1-6H3/t24-,26+,27+,28-,29+,30+,32+,33-/m1/s1

InChI key

FPBODWXATDKICU-FLFWOSPYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Avarzed Amgalanbaatar et al.
Lipids, 50(8), 799-804 (2015-07-01)
This study demonstrated that the cells of Helicobacter felis and Helicobacter cinaedi spontaneously absorb cholesterol added to the medium. A recent study by our group has revealed that phosphatidylethanolamine (PtdEtn) of Helicobacter pylori contains myristic acid as the most predominant

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service