Skip to Content
Merck
All Photos(1)

Key Documents

C6491

Sigma-Aldrich

Chrysomycin B

≥98% (HPLC), solid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H28O9
CAS Number:
Molecular Weight:
496.51
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

solid

solubility

DMF: soluble
DMSO: soluble
ethanol: moderately soluble
methanol: moderately soluble

antibiotic activity spectrum

neoplastics

Mode of action

enzyme | inhibits

storage temp.

2-8°C

SMILES string

COc1cc(C)cc2C(=O)Oc3c(cc(OC)c4c(O)ccc([C@@H]5O[C@H](C)[C@H](O)[C@@](C)(O)[C@H]5O)c34)-c12

InChI

1S/C27H28O9/c1-11-8-15-19(17(9-11)33-4)14-10-18(34-5)21-16(28)7-6-13(20(21)22(14)36-26(15)31)23-25(30)27(3,32)24(29)12(2)35-23/h6-10,12,23-25,28-30,32H,1-5H3/t12-,23+,24+,25+,27-/m1/s1

InChI key

BJPYMDSMDBCKEP-QSMCFSHASA-N

Biochem/physiol Actions

Antibiotic from Streptomyces sp. Inhibits the catalytic activity of human topoisomerase II.
Antibiotic from Streptomyces sp. Inhibits the catalytic activity of human topoisomerase II. Exhibits antitumor activity against human cell lines K562, HT29, MCF7, PC6, and MKN28.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antitumor activity of chrysomycins M and V.
J A Matson et al.
The Journal of antibiotics, 42(9), 1446-1448 (1989-09-01)
Madan K Kharel et al.
Natural product reports, 29(2), 264-325 (2011-12-22)
Covering: 1997 to 2010. The angucycline group is the largest group of type II PKS-engineered natural products, rich in biological activities and chemical scaffolds. This stimulated synthetic creativity and biosynthetic inquisitiveness. The synthetic studies used five different strategies, involving Diels-Alder
N S Burres et al.
Journal of natural products, 55(11), 1582-1587 (1992-11-01)
A simple microtiter assay for the detection of compounds that bind DNA is described. Agents that displace methyl green from DNA are detected spectrophotometrically by a decrease in absorbance at 630 nm. The feasibility of using the assay for detecting
U Weiss et al.
The Journal of antibiotics, 35(9), 1194-1201 (1982-09-01)
The yellow antibiotic chrysomycin, isolated in crystalline form in 1955, is found to consist of two closely related components, a major one, chrysomycin A, and a minor one, chrysomycin B. They differ only through the replacement of a vinyl group

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service