Skip to Content
Merck
All Photos(1)

Key Documents

C5788

Sigma-Aldrich

Complement C5a human

recombinant, expressed in E. coli, ~95% (SDS-PAGE), lyophilized powder

Synonym(s):

C5a anaphylatoxin, rC5a

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
Pricing and availability is not currently available.

biological source

human

Quality Level

recombinant

expressed in E. coli

Assay

~95% (SDS-PAGE)

form

lyophilized powder

technique(s)

activity assay: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... C5(727)

Looking for similar products? Visit Product Comparison Guide

Application

Complement C5a human is an important terminal component of the complement cascade. It is a direct mediator of inflammation, and has been identified as a novel biomarker for pain and inflammation following surgery

Biochem/physiol Actions

A mixture of C5a (~35%) and C5a having an added methionyl residue at the amino terminus (~65%); exhibits biological activities similar to serum-derived C5a. C5a is a (11.2kDa) proteolytic fragment of the C5 α-chain through the action of C5 convertases in the classical and alternative complement pathway (C4b2a4b, C3bBb3b). C5a is an anaphylatoxin. It acts as an inflammatory chemoattractant. C5a stimulation of human neutrophils leads to STAT3 phosphorylation on Ser727. It mediates IL-8 release from bronchial epithelial cells. C5a anaphylatoxin activity on hepatocytes results indirectly from interaction with nonparenchymal cell via prostanoid secretion.
C5a, in addition to being a direct mediator of inflammation, can induce both IL-8 (interleukin-8) synthesis and high levels of IL-8 release from monocytes. This secondary effect serves as an amplification mechanism for inflammation at sites of infection or trauma. C5a exerts its effect through a G-protein coupled receptor, CD88.

Quality

Mol. Wt.: ~8.6 kDa (non-glycosylated, with glutathione attached to cysteine 27).

Other Notes

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Markus Bosmann et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 27(12), 5010-5021 (2013-08-29)
We investigated how complement activation promotes tissue injury and organ dysfunction during acute inflammation. Three models of acute lung injury (ALI) induced by LPS, IgG immune complexes, or C5a were used in C57BL/6 mice, all models requiring availability of both
Andrea L Conroy et al.
Cell host & microbe, 13(2), 215-226 (2013-02-19)
Placental malaria (PM) is a major cause of fetal growth restriction, yet the underlying mechanism is unclear. Complement C5a and C5a receptor levels are increased with PM. C5a is implicated in fetal growth restriction in non-infection-based animal models. In a
Julia Phieler et al.
Journal of immunology (Baltimore, Md. : 1950), 191(8), 4367-4374 (2013-09-18)
Obese adipose tissue (AT) inflammation contributes critically to development of insulin resistance. The complement anaphylatoxin C5a receptor (C5aR) has been implicated in inflammatory processes and as regulator of macrophage activation and polarization. However, the role of C5aR in obesity and
A Conway Morris et al.
British journal of anaesthesia, 111(5), 778-787 (2013-06-13)
Nosocomial infection occurs commonly in intensive care units (ICUs). Although critical illness is associated with immune activation, the prevalence of nosocomial infections suggests concomitant immune suppression. This study examined the temporal occurrence of immune dysfunction across three immune cell types
J David Clark et al.
Anesthesiology, 104(6), 1274-1282 (2006-05-30)
Activation of the complement system is one component of the inflammatory response. Various components of the complement system participate in killing foreign organisms, recruiting immune cells, enhancing edema, and stimulating cytokine formation. Complement-mediated enhancement of the inflammation surrounding surgical incisions

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service