Skip to Content
Merck
All Photos(1)

Key Documents

C5749

Sigma-Aldrich

CGP 52608

≥98%, solid

Synonym(s):

1-[3-Allyl-4-oxo-thiazolidine-2-ylidene]-4-methyl-thiosemicarbazone, N-Methyl-2-[4-oxo-3-(2-propen-1-yl)-2-thiazolidinylidene]-hydrazinecarbothioamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$195.00
25 MG
$762.00

$195.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$195.00
25 MG
$762.00

About This Item

Empirical Formula (Hill Notation):
C8H12N4OS2
CAS Number:
Molecular Weight:
244.34
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$195.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

Assay

≥98%

form

solid

solubility

DMSO: >30 mg/mL
H2O: insoluble

storage temp.

−20°C

SMILES string

CNC(=S)N\N=C1/SCC(=O)N1CC=C

InChI

1S/C8H12N4OS2/c1-3-4-12-6(13)5-15-8(12)11-10-7(14)9-2/h3H,1,4-5H2,2H3,(H2,9,10,14)/b11-8-

InChI key

DDYJDIHOSRTMSE-FLIBITNWSA-N

Application

CGP 52608 has been used as a RAR related orphan receptor α (RORα) synthetic agonist in THP-1 macrophages,[1] in human monocyte cells THP1[2] and somatic testicular cells.[3]

Biochem/physiol Actions

CGP 52608 is a specific activator of retinoic acid receptor-related orphan receptor α (RORA).
CGP 52608 is a thiazolidine derivative[4] and an antiarthritic compound.[5] It possesses immunostimulatory functionality[4] and elicits antiproliferative effect in colon tumors.[6] It is cytotoxic towards neural cell lines.[5] CGP 52608 induces apoptosis in tumor[6] and also favors G0-G1 cell cycle phase accumulation in neural cell lines.[5]

Features and Benefits

This compound is featured on the Nuclear Receptors (Non-Steroids) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrea Balla et al.
Neuropharmacology, 56(5), 915-921 (2009-04-18)
Deficits in N-methyl-D-aspartate receptor (NMDAR)-mediated neurotransmission may underlie dopaminergic hyperactivity in schizophrenia. Dysregulation of the GABAergic system has also been implicated. In this study we investigated a role for GABA(B) receptors as an intermediate step in the pathway leading from
Michal Karasek et al.
Journal of pineal research, 34(4), 294-296 (2003-03-29)
Melatonin exerts a marked antiproliferative action in numerous experimentally-induced tumors in vivo as well as in both animal and human cell lines in vitro. However, the mechanisms of oncostatic action of melatonin is not clear, and the involvement of both
M Karasek et al.
Biological signals and receptors, 8(1-2), 75-78 (1999-03-23)
The antiproliferative effects of melatonin and CGP 52608, an exogenous ligand for RZR/ROR receptors, are compared in the present paper. Both compounds exerted similar inhibitory effects on the proliferation of neoplastic cells in mouse colonic adenocarcinoma, DU 145 human prostate
K Winczyk et al.
Journal of pineal research, 31(2), 179-182 (2001-09-14)
The effects of melatonin and the thiazolinidinedione derivative CGP 52608 on apoptosis of Colon 38 cancer cells were investigated. Male mice were implanted subcutaneously with a suspension of Colon 38 cells. Ten days after induction of tumors, the animals were
Shou-Long Deng et al.
Oncotarget, 8(66), 110592-110605 (2018-01-05)
Because androgen function is regulated by its receptors, androgen-androgen receptor signaling is crucial for regulating spermatogenesis. Androgen is mainly testosterone secreted by testis. Based on the results of early studies in goats, the administration of melatonin over an extended period

Articles

We offer many products related to non-steroid nuclear receptors for your research needs.

Questions

  1. Hi, I wonder how long time I can use it the CGP52608 (C5749) after delivery since there is no expiry date on the CoA ?

    1 answer
    1. This product is not assigned an expiration date or recommended retest date. Products with no expiration date or recommended retest date should be routinely inspected by customers to ensure they perform as expected. These products are also subject to a one year warranty from the date of shipment. For more information, you may access the "Product Dating Information" document under "ADDITIONAL USEFUL DOCUMENTS ABOUT OUR PRODUCTS" at the bottom of the Quality Services page with this link: https://www.sigmaaldrich.com/life-science/quality-and-regulatory-management/quality-services.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service