Skip to Content
Merck
All Photos(1)

Key Documents

C5556

Sigma-Aldrich

Carboxyethyl-γ-aminobutyric acid

Hybri-Max, powder, γ-irradiated, BioXtra, suitable for hybridoma

Synonym(s):

CEGABA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H13NO4
CAS Number:
Molecular Weight:
175.18
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

Hybri-Max

sterility

γ-irradiated

product line

BioXtra

form

powder

technique(s)

cell culture | hybridoma: suitable

impurities

endotoxin, tested

solubility

cell culture medium: 1.5 mg/mL (Store solution at −20°C.)
H2O: soluble (Store solution at −20°C.)

SMILES string

OC(=O)CCCNCCC(O)=O

InChI

1S/C7H13NO4/c9-6(10)2-1-4-8-5-3-7(11)12/h8H,1-5H2,(H,9,10)(H,11,12)

InChI key

SRGQUICKDUQCKO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Stimulates hybridoma growth.

Biochem/physiol Actions

A polyamine derivative that stimulates the growth of hybridoma cells.
Cell growth promoter

Legal Information

Hybri-Max is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service