Skip to Content
Merck
All Photos(1)

Key Documents

C5392

Sigma-Aldrich

6-Cyanopurine

~90%

Synonym(s):

6-Purinecarbonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H3N5
CAS Number:
Molecular Weight:
145.12
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

~90%

form

powder

mp

186-189 °C (lit.)

SMILES string

N#Cc1ncnc2nc[nH]c12

InChI

1S/C6H3N5/c7-1-4-5-6(10-2-8-4)11-3-9-5/h2-3H,(H,8,9,10,11)

InChI key

NHYQQYMSESLQDX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Alice Carvalho et al.
Organic & biomolecular chemistry, 2(7), 1019-1024 (2004-03-23)
An extremely simple method for the selective synthesis of 9-aryl and 9-alkyl 6-alkoxy or 6-alkoxyformimidoylpurines from the corresponding 6-cyanopurines is described. The reaction is carried out with methanol or ethanol in the presence of DBU. At room temperature, nucleophilic attack
J B Peterson et al.
Journal of bacteriology, 151(3), 1473-1484 (1982-09-01)
A soluble aldehyde dehydrogenase (EC 1.2.1.3) was partially purified from Rhizobium japonicum bacteroids and from free-living R. japonicum 61A76. The enzyme was activated by NAD+, NADH, and dithiothreitol, and it reduced NAD(P)+. Acetaldehyde, propionaldehyde, butyraldehyde, benzaldehyde, and succinic semialdehyde were
Zadkiel Alvarez et al.
Antimicrobial agents and chemotherapy, 54(12), 5329-5336 (2010-10-06)
Bacillus anthracis, the etiological agent of anthrax, has a dormant stage in its life cycle known as the endospore. When conditions become favorable, spores germinate and transform into vegetative bacteria. In inhalational anthrax, the most fatal manifestation of the disease

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service