Skip to Content
Merck

Skip To

C5233

Carboxypeptidase B from human pancreas

50-55 units/mg protein carboxypeptidase B

Synonym(s):

Peptidyl-L-Lysine[L-arginine] hydrolase

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.54
EC Number:
MDL number:
Specific activity:
50-55 units/mg protein carboxypeptidase B
Biological source:
human pancreas
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

human pancreas

Quality Segment

form

solution

specific activity

50-55 units/mg protein carboxypeptidase B

impurities

≤0.2% chymotrypsin, ≤0.2% trypsin, ≤1 unit/mg protein carboxypeptidase A

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

SMILES string

S(Cc3ccccc3)C(=O)C(NC(=O)C2N(CCC2)C(=O)C(NC(=O)C(NC(=O)CCC(=O)O)C)C)Cc1ccccc1

InChI

1S/C31H38N4O7S/c1-20(32-26(36)15-16-27(37)38)28(39)33-21(2)30(41)35-17-9-14-25(35)29(40)34-24(18-22-10-5-3-6-11-22)31(42)43-19-23-12-7-4-8-13-23/h3-8,10-13,20-21,24-25H,9,14-19H2,1-2H3,(H,32,36)(H,33,39)(H,34,40)(H,37,38)

InChI key

TWURVFFNODFJBJ-UHFFFAOYSA-N

Gene Information

human ... CPB1(1360)

General description

Carboxypeptidase B is mapped to human chromosome 3q24.[1] Carboxypeptidase B belongs to A/B subfamily of carboxypeptidases.

Application

Carboxypeptidase B from Sigma has been used as a reference for assaying carboxypeptidase activity in lysed pituitary granules derived from the anterior and intermediate lobes of rat.[2] The enzyme has also been used to digest plasma samples by removing C-terminal basic amino acids, to get a distinct band for each allotype during C4 electrophoresis.[3]

Biochem/physiol Actions

Carboxypeptidase B (or peptidyl-L-lysine (-L-arginine) hydrolase) catalyzes the hydrolysis of the basic amino acids, lysine, arginine, and ornithine from the C-terminal position of polypeptides. It has been shown to be a single polypeptide of 34,000 Da. Trypsin is capable of converting native enzyme to the active enzyme, carboxypeptidase B II in vitro.[4] The optimum pH is found to be 9.0. The enzyme may be used for sequence analysis by successive cleavage of C-terminal basic amino acids. It can also be used as a serum marker for the diagnosis of acute pancreatitis.
Mutations in the carboxypeptidase B (CPB1) gene is implicated with increased susceptibility to pancreatic cancer development and progression. Elevated levels of CPB1 is associated with low grade breast tumors and lymph node positive grade 1 tumors.

Physical form

Solution in 0.05 M NaOAc pH 5.0 + 1.0 M NaCl + 0.01% NaN3

Other Notes

One unit will hydrolyze 1 μmole of hippuryl-L-arginine per minute at pH 7.7 at 25 °C


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

  1. I have a question about the Carboxypeptidase B (C-5233). Generally, how much volume of buffer solution (0.05M NaOAc pH 5.0+1.0M NaCl+0.01% NaN3) to add to make the stock? What is the storage temp. for the stock solution after reconstitution? Thank you!

    1 answer
    1. This product is provided as a solution with a minimum concentration of 0.50 mg protein/mL Carboxypeptidase B in 0.05 M NaOAc pH 5.0, containing 1.0 M NaCl and 0.01% NaN3. The concentration is lot-specific and reported in the product Certificate of Analysis. The maximum volume, based on the product specification, is 0.2 mL. This product is stored at −20°C and ships on dry ice. Please see the link below to review a sample or lot-specific Certificate of Analysis:
      https://www.sigmaaldrich.com/product/sigma/c5233#product-information

      Helpful?

Reviews

No rating value

Active Filters