C5108
Cathepsin G from human sputum
≥0.2 units/mg protein, salt-free, lyophilized powder
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
CAS Number:
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.32
Recommended Products
biological source
human sputum
form
salt-free, lyophilized powder
specific activity
≥0.2 units/mg protein
UniProt accession no.
shipped in
wet ice
storage temp.
2-8°C
Gene Information
human ... CTSG(1511)
Biochem/physiol Actions
Cathepsin G has pro-apoptotic activity and can activate caspases in vitro.
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Prasad Tongaonkar et al.
PloS one, 7(3), e32469-e32469 (2012-03-27)
The azurophilic granules of human neutrophils contain four α-defensins called human neutrophil peptides (HNPs 1-4). HNPs are tridisulfide-linked antimicrobial peptides involved in the intracellular killing of organisms phagocytosed by neutrophils. The peptides are produced as inactive precursors (proHNPs) which are
E D Son et al.
Dermatology (Basel, Switzerland), 224(4), 352-360 (2012-07-05)
Cathepsin G, a serine protease that is activated by ultraviolet (UV) radiation, increases matrix metalloproteinase-1 (MMP-1) expression in fibroblasts through fibronectin (Fn) fragmentation and promotes the conversion of proMMP-1 to active MMP-1. This study investigated whether [2-[3-[[(1-benzoyl-4-piperidinyl)methylamino]carbonyl]-2-naphthalenyl]-1-(1-naphthalenyl)-2-oxoethyl]-phosphonic acid (KPA), a
Alice V Dubois et al.
American journal of respiratory cell and molecular biology, 47(1), 80-86 (2012-02-22)
Uncontrolled proteolysis by neutrophil serine proteases (NSPs) in lung secretions is a hallmark of cystic fibrosis (CF). We have shown that the active neutrophil elastase, protease 3, and cathepsin G in CF sputum resist inhibition in part by exogenous protease
Jun Zhang et al.
American journal of physiology. Renal physiology, 303(4), F593-F603 (2012-06-08)
The renin-angiotensin system (RAS) is well studied for its regulation of blood pressure and fluid homeostasis, as well as for increased activity associated with a variety of diseases and conditions, including cardiovascular disease, diabetes, and kidney disease. The enzyme renin
Dawid Dębowski et al.
Peptides, 35(2), 276-284 (2012-04-21)
A series of linear and cyclic fragments and analogs of two peptides (OGTI and HV-BBI) isolated from skin secretions of frogs were synthesized by the solid-phase method. Their inhibitory activity against several serine proteinases: bovine β-trypsin, bovine α-chymotypsin, human leukocyte
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service