Skip to Content
Merck
All Photos(1)

Key Documents

C4540

Sigma-Aldrich

Citric acid

suitable for plant cell culture

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC(COOH)(CH2COOH)2
CAS Number:
Molecular Weight:
192.12
Beilstein:
782061
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:

expl. lim.

8 %, 65 °F

technique(s)

cell culture | plant: suitable

pKa 

(1) 3.13, (2) 4.76, (3) 6.4

mp

153-159 °C (lit.)

SMILES string

OC(=O)CC(O)(CC(O)=O)C(O)=O

InChI

1S/C6H8O7/c7-3(8)1-6(13,5(11)12)2-4(9)10/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12)

InChI key

KRKNYBCHXYNGOX-UHFFFAOYSA-N

Gene Information

human ... SRC(6714)

Looking for similar products? Visit Product Comparison Guide

Application

In addition to its function as an iron chelator, citrate can support cholesterol/sterol, ubiquinone and isoprenoid biosynthesis in cell culture applications.

replaced by

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maria José Garcia-Fernandez et al.
Biofouling, 29(3), 261-271 (2013-02-27)
Prevention and management of wound infections receive a lot of attention, since the presence of micro-organisms interferes with the wound-healing process. The aim of this work was to use cyclodextrins (CDs) to endow hydrogels and gauzes with the ability to
Shihhui Huang et al.
Hippocampus, 25(8), 963-975 (2015-01-24)
Work on laboratory and wild rodents suggests that domestication may impact on the extent of adult hippocampal neurogenesis and its responsiveness to regulatory factors. There is, however, no model of laboratory rodents and their nondomesticated conspecifics that would allow a
Konrad Hoetzenecker et al.
European heart journal, 36(11), 676-685 (2013-01-17)
Supernatants of serum-free cultured mononuclear cells (MNC) contain a mix of immunomodulating factors (secretome), which have been shown to attenuate detrimental inflammatory responses following myocardial ischaemia. Inflammatory dilated cardiomyopathy (iDCM) is a common cause of heart failure in young patients.
Wayne B Batchelor et al.
Circulation, 106(12), 1470-1476 (2002-09-18)
The relative anti-aggregatory effects of currently prescribed platelet glycoprotein IIb/IIIa receptor antagonists during and after percutaneous coronary intervention for acute coronary syndromes have not been established. We randomized 70 acute coronary syndrome patients undergoing percutaneous coronary intervention to receive abciximab
Benjamin Nota et al.
American journal of human genetics, 92(4), 627-631 (2013-04-09)
The Krebs cycle is of fundamental importance for the generation of the energetic and molecular needs of both prokaryotic and eukaryotic cells. Both enantiomers of metabolite 2-hydroxyglutarate are directly linked to this pivotal biochemical pathway and are found elevated not

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service