Skip to Content
Merck
All Photos(1)

Key Documents

C4351

Supelco

CoZap for Coomassie Blue removal

Sign Into View Organizational & Contract Pricing

Select a Size

25 EA
$222.00

$222.00


Estimated to ship onMay 23, 2025



Select a Size

Change View
25 EA
$222.00

About This Item

UNSPSC Code:
12352200
NACRES:
NB.51

$222.00


Estimated to ship onMay 23, 2025


availability

not available in EU

technique(s)

protein staining: suitable

Application

CoZap is used for rapid removal of Coomasie blue stain. The unique CoZap pad is placed directly into your destaining tray and abosrbs all free dye in solution. This eliminates the need to change the destaining solution.

Features and Benefits

Decrease destaining time by 20%
No charcol or dye residues
One pad can destain up to 10 gels
No need to change destaining solution

Legal Information

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhongyu Li et al.
BMC microbiology, 8, 90-90 (2008-06-11)
C. trachomatis organisms carry a cryptic plasmid that encodes 8 open reading frames designated as pORF1 to 8. It is not clear whether all 8 pORFs are expressed during C. trachomatis infection in humans and information on the functionality of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service