Skip to Content
Merck
All Photos(1)

Key Documents

C4170

Sigma-Aldrich

Chondroitin disaccharide Δdi-6S sodium salt

Synonym(s):

α-ΔUA-[1→3]-GalNAc-6S

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C14H19NO14SNa2
CAS Number:
Molecular Weight:
503.34
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:

storage temp.

−20°C

SMILES string

[Na+].[Na+].CC(=O)N[C@H]1C(O)O[C@H](COS([O-])(=O)=O)[C@H](O)[C@@H]1OC2OC(=C[C@H](O)[C@H]2O)C([O-])=O

Application

Chondroitin, a glucosaminoglycan (GAG), is a polysaccharide chain of alternating units of N-acetylgalactosamine (GalNAc) and glucuronic acid (GlcA) that can be sulfated on the either or both GalNAc and GlcA units. Chondroitin and its sulfates are frequently attached to proteins to form proteoglycans.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

ΔUA = 4-deoxy-L-threo-hex-4-enopyranosyluronic acid; GalNAc = N-acetyl-D-galactosamine; 6S = 6-sulfate

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Glucosamine and chondroitin sulfate.
Miller KL, Clegg DO.
Rheumatic Diseases Clinics of North America, 23(1), 103-118 (2012)
J C F Kwok et al.
The international journal of biochemistry & cell biology, 44(4), 582-586 (2012-01-24)
Chondroitin sulfate is a glycosaminoglycan composed of N-acetylgalactosamine and glucuronic acid. It attaches to a core protein to form chondroitin sulfate proteoglycan (CSPG). Being a major component of the brain extracellular matrix, CSPGs are involved in neural development, axon pathfinding
Yulin Li et al.
Chemical Society reviews, 41(6), 2193-2221 (2011-11-26)
Injectable hydrogels with biodegradability have in situ formability which in vitro/in vivo allows an effective and homogeneous encapsulation of drugs/cells, and convenient in vivo surgical operation in a minimally invasive way, causing smaller scar size and less pain for patients.
Juliane Salbach et al.
Journal of molecular medicine (Berlin, Germany), 90(6), 625-635 (2011-12-22)
To meet the growing need for tissue replacement materials for our aging population, the development of new adaptive biomaterials is essential. The tissues with the highest demand for implant materials are skin and bone. These tissues share various similarities, including
The role of chondroitin sulfate proteoglycans in regeneration and plasticity in the central nervous system.
Galtrey CM, Fawcett JW.
Brain Research. Brain Research Reviews, 54, 1-18 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service