Skip to Content
Merck
All Photos(1)

Key Documents

C-39365

Sigma-Aldrich

Skeletal Muscle Cell Growth Medium

SupplementMix containing all media supplements pre-mixed in one vial, 1 Mix for 500 ml

Synonym(s):

Recommended with C-23260

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.71
Pricing and availability is not currently available.

form

aqueous solution

packaging

pkg of 1 vial

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−20°C

General description

Skeletal Muscle Cell Growth Medium SupplementMix (basal medium sold separately)
To form a complete medium, it should be combined with the basal medium: C-23260

Application

Skeletal Muscle Cell Growth Medium is a low-serum medium (5% V/V) developed for the expansion of human skeletal muscle cell cultures. The media are optimized for primary human cells, but can also be used for murine, rat, canine, and rabbit skeletal muscle cells.Recommended for:Human Skeletal Muscle Cells (SkMC)

Components

Part of a Cell Culture System offered in a variety of formats:Growth Medium (Ready-to-use): Includes Basal Medium and SupplementMixGrowth Medium Kit: Includes Basal Medium and SupplementPackSupplementMix: Contains all media supplements premixed in one vialSupplementPack: Contains all media supplements as individual vials

Quality

All lots of PromoCell Skeletal Muscle Cell Growth Medium are subjected to comprehensive quality control tests using human primary SkMC. Each lot is checked for growth promoting activity, adherence rate, and typical morphology of the tested SkMC. Approved in-house lots of media are used as a reference. In addition, all lots of media have been tested for the absence of microbial contaminants (fungi, bacteria, mycoplasma).

Preparation Note

Final supplement concentrations after addition to the basal medium:Fetal Calf Serum: 0.05 ml / mlFetuin (bovine): 50 µg / mlEpidermal Growth Factor (recombinant human): 10 ng / mlBasic Fibroblast Growth Factor (recombinant human): 1 ng / mlInsulin (recombinant human): 10 µg / mlDexamethasone: 0.4 µg / ml

Storage and Stability

Store the SupplementMix at -20°C in the dark immediately after arrival. If stored properly, it is stable until the expiry date stated on the label. After adding the supplements to the Basal Medium, the shelf life of the complete medium is 6 weeks at 4 to 8°C. Do not freeze the complete medium. For use, pre-warm only an aliquot of the complete medium and keep the remaining medium refrigerated at 4 to 8°C.

Subculture Routine

Click here for more information.

recommended

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Primary human skeletal muscle cells and culture medium optimized for the in vitro cultivation and differentiation of primary human skeletal muscle cells. Protocols for handling, cell passaging, media, and product use.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service