Skip to Content
Merck
All Photos(4)

Key Documents

C3635

Sigma-Aldrich

4-Chloro-DL-phenylalanine methyl ester hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$137.00
5 G
$493.00
25 G
$1,870.00

$137.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$137.00
5 G
$493.00
25 G
$1,870.00

About This Item

Linear Formula:
ClC6H4CH2CH(NH2)CO2CH3·HCl
CAS Number:
Molecular Weight:
250.12
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.32

$137.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

powder

Quality Level

mp

186-189 °C (lit.)

storage temp.

−20°C

SMILES string

Cl.COC(=O)C(N)Cc1ccc(Cl)cc1

InChI

1S/C10H12ClNO2.ClH/c1-14-10(13)9(12)6-7-2-4-8(11)5-3-7;/h2-5,9H,6,12H2,1H3;1H

InChI key

GCBCWTWQAFLKJG-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

4-Chloro-DL-phenylalanine methyl ester hydrochloride stimulates glucose intolerance in pregnant mice.[1] Studies have shown that 4-Chloro-DL-phenylalanine methyl ester hydrochloride is associated with cognitive defects in rodents.[2]

Application

4-Chloro-DL-phenylalanine methyl ester hydrochloride has been used to induce 5-hydroxytryptamine (5-HT) depletion in rats.[3]

Biochem/physiol Actions

Tryptophan hydroxylase inhibitor.
Tryptophan hydroxylase inhibitor. Crosses blood brain barrier better than p−chlorophenylalanine.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jessica E Monk et al.
Frontiers in behavioral neuroscience, 12, 246-246 (2018-11-09)
Negative affective states such as anxiety and depression pose a risk to animal welfare, however, practical tests for assessing these states in animals are limited. In humans, anxious individuals are shown to pay more attention toward threatening information than non-anxious
Vortioxetine dose-dependently reverses 5-HT depletion-induced deficits in spatial working and object recognition memory: a potential role for 5-HT1A receptor agonism and 5-HT3 receptor antagonism
du Jardin KG, et al.
European Neuropsychopharmacology, 24(1), 160-171 (2014)
Serotonin regulates pancreatic beta cell mass during pregnancy
Kim H, et al.
Nature Medicine, 16(7), 804-804 (2010)
Serotonin depletion results in a decrease of the neuronal activation caused by rivastigmine in the rat hippocampus
Kornum BR, et al.
Brain Research, 1073, 262-268 (2006)
Lucielli Savegnago et al.
Progress in neuro-psychopharmacology & biological psychiatry, 31(6), 1261-1269 (2007-06-26)
In this study, the antidepressant-like effect caused by diphenyl diselenide on rat forced swimming test (FST) was investigated. The involvement of the monoaminergic system in the antidepressant-like effect was also evaluated. Diphenyl diselenide (0.1-30 mg/kg), given by oral route (p.o.)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service