C-28057
Macrophage Base Medium DXF
Basal Medium, 250 ml
Synonym(s):
Macrophage Culture Medium
About This Item
Recommended Products
form
liquid
packaging
pkg of 250 ml
technique(s)
cell culture | mammalian: suitable
shipped in
wet ice
storage temp.
2-8°C
General description
Application
Components
Quality
Storage and Stability
Subculture Routine
Storage Class Code
10 - Combustible liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Explore over 350 PromoCell products on Sigma.com, including cryopreserved blood cells and cell culture media for various human cells.
Protocols
Generate phagocytic M1 and M2 macrophages from human PBMCs in 10 days in serum-free and xeno-free cell culture media. Explore over 350 PromoCell products.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service