C2797
4-(Chloromethyl)benzoic acid
Synonym(s):
α-Chloro-p-toluic acid, α-Chloro-p-toluylic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C8H7ClO2
CAS Number:
Molecular Weight:
170.59
Beilstein:
1907970
EC Number:
MDL number:
UNSPSC Code:
12352100
Recommended Products
mp
201-202 °C (lit.)
storage temp.
2-8°C
SMILES string
OC(=O)c1ccc(CCl)cc1
InChI
1S/C8H7ClO2/c9-5-6-1-3-7(4-2-6)8(10)11/h1-4H,5H2,(H,10,11)
InChI key
OITNBJHJJGMFBN-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
replaced by
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service