Skip to Content
Merck
All Photos(1)

Key Documents

C2749

Sigma-Aldrich

Cystatin C human

recombinant, expressed in Pichia pastoris

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202

recombinant

expressed in Pichia pastoris

Quality Level

form

solid

shipped in

wet ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Physical form

Lyophilized from ammonium bicarbonate

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hyun Jung Chung et al.
PloS one, 10(7), e0133417-e0133417 (2015-07-18)
The high incidence of acute and chronic kidney injury due to various environmental factors such as heavy metals or chemicals has been a major problem in developing countries. However, the diagnosis of kidney injury in these areas can be more
Olgica Trenchevska et al.
PloS one, 9(6), e100713-e100713 (2014-06-24)
Human protein diversity arises as a result of alternative splicing, single nucleotide polymorphisms (SNPs) and posttranslational modifications. Because of these processes, each protein can exists as multiple variants in vivo. Tailored strategies are needed to study these protein variants and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service