Skip to Content
Merck
All Photos(1)

Documents

C2639

Sigma-Aldrich

Cell Freezing Medium-Serum-free 1×

sterile-filtered, suitable for cell culture

Synonym(s):

Cryopreservation Medium, Serum-Free Freezing Medium

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.75

sterility

sterile-filtered

Quality Level

form

liquid

technique(s)

cell culture | mammalian: suitable
cryopreservation: suitable

shipped in

ambient

storage temp.

2-8°C

Application

Cell Freezing Medium-Serum-free 1× has been used as a cryopreservation medium for somatic cell sampling and recovery (SCSR) exfoliated cells cultured in growth media and dihydrofolate reductase (DHFR)-deficient Chinese hamster ovary (CHO) cells grown in serum-free growth medium.
Sigma′s Cell Freezing Medium, Serum-free is a cryopreservation medium which does not contain DMSO or serum.

Other Notes

Ready-to-use formula that does not contain dimethyl sulfoxide or serum.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jeong Soo Park et al.
Protein expression and purification, 70(2), 231-235 (2009-10-13)
Hepatocyte growth factor (HGF) is a potent multi-functional protein that affects morphogenesis, cell migration, organ regeneration, and tumor invasion in various tissues, and has thus been considered to have potential as a therapeutic target in various diseases. In our current
Identification of a native novel oncolytic immunoglobulin on exfoliated colon epithelial cells: A bispecific heterodimeric chimera of IgA/IgG
Albaugh G P, et al.
Open Medicine : A Peer-Reviewed, Independent, Open-Access Journal, 10(6), 126-150 (2020)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service