Skip to Content
Merck
All Photos(1)

Documents

C2335

Sigma-Aldrich

Casein Kinase I substrate

≥95% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C85H139N27O35S
CAS Number:
Molecular Weight:
2131.24
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

Assay

≥95% (HPLC)

form

powder

potency

172 μM Km

composition

Peptide content, ~70%

UniProt accession no.

storage temp.

−20°C

SMILES string

CC[C@H](C)[C@H](NC(=O)[C@H](CO)NC(=O)[C@H](CCSC)NC(=O)[C@H](C)NC(=O)[C@H](CCC(O)=O)NC(=O)[C@H](CC(O)=O)NC(=O)[C@H](CCC(O)=O)NC(=O)[C@H](CCC(O)=O)NC(=O)[C@H](CC(O)=O)NC(=O)[C@H](CC(O)=O)NC(=O)[C@H](Cc1c[nH]cn1)NC(=O)[C@H](CC(C)C)NC(=O)[C@H](CC(O)=O)NC(=O)[C@H](CCCCN)NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@@H](N)CCCNC(N)=N)C(=O)N[C@@H]([C@@H](C)O)C(=O)N[C@@H](C)C(O)=O

InChI

1S/C85H139N27O35S/c1-9-38(4)64(81(144)112-65(41(7)114)82(145)97-40(6)83(146)147)111-80(143)56(35-113)110-73(136)49(23-27-148-8)98-66(129)39(5)96-68(131)46(17-20-57(115)116)102-76(139)52(30-60(121)122)107-72(135)48(19-22-59(119)120)101-71(134)47(18-21-58(117)118)103-77(140)53(31-61(123)124)109-79(142)55(33-63(127)128)108-75(138)51(29-42-34-92-36-95-42)105-74(137)50(28-37(2)3)104-78(141)54(32-62(125)126)106-70(133)44(15-10-11-24-86)100-69(132)45(16-13-26-94-85(90)91)99-67(130)43(87)14-12-25-93-84(88)89/h34,36-41,43-56,64-65,113-114H,9-33,35,86-87H2,1-8H3,(H,92,95)(H,96,131)(H,97,145)(H,98,129)(H,99,130)(H,100,132)(H,101,134)(H,102,139)(H,103,140)(H,104,141)(H,105,137)(H,106,133)(H,107,135)(H,108,138)(H,109,142)(H,110,136)(H,111,143)(H,112,144)(H,115,116)(H,117,118)(H,119,120)(H,121,122)(H,123,124)(H,125,126)(H,127,128)(H,146,147)(H4,88,89,93)(H4,90,91,94)/t38-,39-,40-,41+,43-,44-,45-,46-,47-,48-,49-,50-,51-,52-,53-,54-,55-,56-,64-,65-/m0/s1

InChI key

GPNUIDBXUOTUMT-LVTIADEOSA-N

Gene Information

Amino Acid Sequence

Arg-Arg-Lys-Asp-Leu-His-Asp-Asp-Glu-Glu-Asp-Glu-Ala-Met-Ser-Ile-Thr-Ala

Specificity

Vmax with casein kinase-1 is 6-fold higher than for casein. Rate of phosphorylation by casein kinase-2 is negligible.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Avanti Gokhale et al.
The Journal of cell biology, 186(6), 817-824 (2009-09-16)
Experimental analysis of isolated ciliary/flagellar axonemes has implicated the protein kinase casein kinase I (CK1) in regulation of dynein. To test this hypothesis, we developed a novel in vitro reconstitution approach using purified recombinant Chlamydomonas reinhardtii CK1, together with CK1-depleted
O Marin et al.
Biochemical and biophysical research communications, 198(3), 898-905 (1994-02-15)
The available information about the specificity determinants of casein kinases-1 and -2 (CK1 and CK2) has been utilized to obtain two new peptide substrates optimally suited for the specific monitoring of these two pleiotropic enzymes. The best substrate developed for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service