The majority of our basal media have a shelf life of 9 months from the date of manufacture when stored between 4°C – 8°C. The shelf life of the supplements is 12 months from the date of manufacture when stored at -20°C. The exact expiry date is specified on the product label.
C-23025
Fibroblast Growth Medium 3
Ready-to-use kit including Basal Medium and SupplementMix, 500 ml
Synonym(s):
Primary Cell Culture Media
About This Item
Recommended Products
form
liquid
packaging
pkg of 500 ml
technique(s)
cell culture | mammalian: suitable
shipped in
wet ice
General description
Application
Components
Quality
Preparation Note
Storage and Stability
Subculture Routine
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Primary human fibroblasts and growth medium for the in vitro cultivation of juvenile and adult fibroblasts. Protocols for handling, cell passaging, media, and product use.
-
Could you please provide the shelf life of Product No. C-23025, PromoCell FIBROBLAST GROWTH MEDIUM 3?
1 answer-
Helpful?
-
-
I will be using cardiac fibroblasts and aside from normal growth media (full amount of Serum) I have to serum starve the cells before my experiments. What media can I purchase where I can only change the amount of serum, not other supplements?
1 answer-
This product does contain serum in the formulation. A pre-formulated supplement is included with the medium that contains serum, bFGF, and insulin. The supplement, as well as any additional serum, may be added or withheld at the end user's discretion. Product 116F-500, an alternative fibroblast growth medium that does not contain serum that may be suitable for serum-starved experimental conditions, may also be of interest. Please see the link below to review this product option:
https://www.sigmaaldrich.com/product/sigma/116f500Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service