Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
5 mg | Check Cart for Availability | $115.00 |
25 mg | Check Cart for Availability | $317.00 |
100 mg | Check Cart for Availability | $1,010.00 |
About This Item
NACRES:
NA.77
UNSPSC Code:
12352211
Assay:
≥99%
Form:
powder
Biochem/physiol Actions
Involved in the action of phospholipase C on sphingomyelin.
Features and Benefits
This compound is featured on the Phospholipase D page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
Other Notes
Contains primarily stearic and nervonic acids.
1 of 1
This Item | |||
|---|---|---|---|
| assay ≥99% | assay ~95% | assay ≥98% (TLC) | assay ≥97% (TLC) |
| form powder | form powder | form lyophilized powder | form powder |
| Quality Level 200 | Quality Level 200 | Quality Level 200 | Quality Level 200 |
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. −20°C |
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



