This product has a polyethylene filter matrix support with a pore size of 10-15 µm.
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
200 ea | Check Cart for Availability | $402.00 |
About This Item
NACRES:
SB.52
UNSPSC Code:
23151816
material
polypropylene
technique(s)
LPLC: suitable
Overall H
5 in.
volume
10 mL
General description
- Precision molded from premium-grade, chemically inert plastic for organic solvents
- Includes reservoir & end caps
- Polyethylene filter matrix support in medium porosity
Application
For use in general chromatographic work for extraction of components, with special applications in isoenzyme and protein studies, hemoglobin fractionation and desalting.
1 of 1
This Item | |||
|---|---|---|---|
| technique(s) LPLC: suitable | technique(s) - | technique(s) - | technique(s) - |
| material polypropylene | material polyethylene | material copper | material polypropylene |
| volume 10 mL | volume 6.5 mL | volume - | volume 6.5 mL |
| Overall H 5 in. | Overall H - | Overall H - | Overall H - |
Still not finding the right product?
Explore all of our products under Chromatography columns, general-purpose
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
We would like to order the Chromatography columns (C2103), but first we would like to know what is the filter pore size?
1 answer-
Helpful?
-


