Skip to Content
Merck
All Photos(1)

Key Documents

C159

Sigma-Aldrich

Calphostin C from Cladosporium cladosporioides

Synonym(s):

UCN-1028c

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H38O14
CAS Number:
Molecular Weight:
790.76
MDL number:
UNSPSC Code:
12352204

storage temp.

−20°C

SMILES string

COc1cc(O)c2C(=O)C(OC)=C(CC(C)OC(=O)Oc3ccc(O)cc3)c4c5C(CC(C)OC(=O)c6ccccc6)=C(OC)C(=O)c7c(O)cc(OC)c(c57)c1c24

InChI

1S/C44H38O14/c1-20(56-43(50)22-10-8-7-9-11-22)16-25-31-32-26(17-21(2)57-44(51)58-24-14-12-23(45)13-15-24)42(55-6)40(49)34-28(47)19-30(53-4)36(38(32)34)35-29(52-3)18-27(46)33(37(31)35)39(48)41(25)54-5/h7-15,18-21,45-47H,16-17H2,1-6H3

InChI key

SRJYZPCBWDVSGO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Highly specific inhibitor of protein kinase C (IC50 = 50 nM); requires light for activation. At higher concentrations (100 nM or greater), it has been shown to inhibit cell proliferation and induce apoptosis in vitro.

replaced by

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yukinori Kozuma et al.
British journal of haematology, 152(5), 631-639 (2011-01-19)
Thrombosis in paroxysmal nocturnal haemoglobinuria (PNH) has been suggested to be due to several pathophysiological states: a suppressed fibrinolytic system, increased leucocyte-derived tissue factor, complement (C')-mediated damage to platelets and endothelia, or increased platelet- and endothelium-derived microparticles (MPs). Because haemolytic
Yuko Abe et al.
Drug metabolism and pharmacokinetics, 26(3), 256-265 (2011-02-15)
UDP-glucuronosyltransferases (UGTs) catalyze the glucuronidation of a wide variety of xeno/endobiotics. Previous studies have reported that human UGT enzymes are phosphorylated and that treatment of cells with protein kinase C (PKC) inhibitors results in decreased UGT activities without affecting the
Peng Sun et al.
American journal of physiology. Renal physiology, 302(6), F679-F687 (2011-12-16)
We examined the effect of angiotensin II (ANG II) on epithelial Na(+) channel (ENaC) in the rat cortical collecting duct (CCD) with single-channel and the perforated whole cell patch-clamp recording. Application of 50 nM ANG II increased ENaC activity, defined
S Lemoine et al.
British journal of anaesthesia, 107(4), 510-518 (2011-08-25)
Desflurane triggers post-conditioning in the diabetic human myocardium. We determined whether protein kinase C (PKC), mitochondrial adenosine triphosphate-sensitive potassium (mitoK(ATP)) channels, Akt, and glycogen synthase kinase-3β (GSK-3β) were involved in the in vitro desflurane-induced post-conditioning of human myocardium from patients
Yanli Hu et al.
European journal of pharmacology, 680(1-3), 49-54 (2012-02-15)
Potassium 2-(1-hydroxypentyl)-benzoate (dl-PHPB) has been shown to have potent neuroprotective effects, such as reducing the infarct volume and improving neurobehavioral deficits in the transient focal cerebral ischemic rat model. The present study is to evaluate the neuroprotective effect of dl-PHPB

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service