Skip to Content
Merck
All Photos(1)

Documents

C1413

Sigma-Aldrich

Complement C7 deficient serum human

for complement assays

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.61

biological source

human

Quality Level

form

solution

technique(s)

activity assay: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... C7(730)

Application

Complement C7 deficiencies in humans are rare, but often associated with recurrent infections by Neisseria spp. (such as meningitis). C7 deficiencies in patients with meningococcal meningitis have shown a mutation which results in an 11 bp deletion in exon 6 resulting in a premature stop codon. Additionally, research has suggested that screening of patients with systemic neisserial infection by CH50 or the APH-50 assay can reveal a C7 deficiency.

Physical form

Supplied as a solution in PBS, pH 7.4

Analysis Note

C7 is depleted by immunoadsorption as judged by a highly sensitive hemolytic assay.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sonia Barroso et al.
Immunology, 118(2), 257-260 (2006-06-15)
Different genetic mutations have been described in complement components resulting in total or subtotal deficiency states. In this work we report the genetic basis of C7 deficiency in a previously reported Spanish patient exhibiting a combined total deficiency of C7
Cryo-EM structures of Trypanosoma brucei gambiense ISG65 with human complement C3 and C3b and their roles in alternative pathway restriction.
S??lzen, et al.
Nature Communications, 14, 2403-2403 (2023)
W P Kolb et al.
Journal of immunology (Baltimore, Md. : 1950), 122(5), 2103-2111 (1979-05-01)
C1q, a subcomponent of the first component of complement, has been isolated from human serum in fully hemolytically active form by affinity column chromatography and gel filtration with Bio-Gel A-5M. The affinity column was prepared by covalent coupling of purified

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service