Skip to Content
Merck
All Photos(1)

Key Documents

C1249

Sigma-Aldrich

CDK4/CyclinD3, active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

CCND3, CMM3, MGC14458, PSK-J3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
51111800
NACRES:
NA.32
Pricing and availability is not currently available.

recombinant

expressed in baculovirus infected Sf9 cells

Quality Level

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

13-19 nmol/min·mg

mol wt

CDK4 subunit ~58 kDa
cyclin D3 ~58 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

Biochem/physiol Actions

CDK4 is a member of the cyclin-dependent protein kinase family and is involved in the control of cell proliferation during the G1 phase of cell cycle. CDK4 forms a complex with the D-type cyclins and is inhibited by p16 (cyclin-dependent kinase inhibitor-2). CDK4 can mediate phosphorylation of the C-terminal region of Rb protein leading to an active transcriptional repression of E2F complex. CDC37 and HSP90 can preferentially associate with the fraction of CDK4 not bound to D-type cyclins. SMAD3 is a major physiologic substrate of the G1 cyclin-dependent kinases CDK4 and CDK2.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.25 mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isao Matsuura et al.
Nature, 430(6996), 226-231 (2004-07-09)
Transforming growth factor-beta (TGF-beta) potently inhibits cell cycle progression at the G1 phase. Smad3 has a key function in mediating the TGF-beta growth-inhibitory response. Here we show that Smad3 is a major physiological substrate of the G1 cyclin-dependent kinases CDK4
J W Harbour et al.
Cell, 98(6), 859-869 (1999-09-28)
We present evidence that phosphorylation of the C-terminal region of Rb by Cdk4/6 initiates successive intramolecular interactions between the C-terminal region and the central pocket. The initial interaction displaces histone deacetylase from the pocket, blocking active transcriptional repression by Rb.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service