Skip to Content
Merck
All Photos(1)

Key Documents

C121

Sigma-Aldrich

7-Chlorokynurenic acid

solid

Synonym(s):

7-Chloro-4-hydroxyquinoline-2-carboxylic acid, 7-Cl-KYNA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H6ClNO3
CAS Number:
Molecular Weight:
223.61
EC Number:
MDL number:
UNSPSC Code:
12352200

form

solid

color

tan

solubility

0.1 M NaOH: 13.7 mg/mL
DMSO: 14 mg/mL

SMILES string

OC(=O)c1cc(O)c2ccc(Cl)cc2n1

Gene Information

human ... GRIN1(2902)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Potent NMDA glutamate receptor antagonist; antagonizes the strychnine-insensitive glycine site of the NMDA receptor.
Potent NMDA glutamate receptor antagonist; antagonizes the strychnine-insensitive glycine site of the NMDA receptor; prevents neurodegeneration produced by quinolinic acid.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service