Skip to Content
Merck
All Photos(1)

Key Documents

C0937

Color Coders for Nalgene® cryogenic vials

green

Synonym(s):

vial cap inserts, vial identifier

Sign Into View Organizational & Contract Pricing

Select a Size

100 EA
$38.40

$38.40


Estimated to ship onMay 19, 2025



Select a Size

Change View
100 EA
$38.40

About This Item

UNSPSC Code:
41103032
NACRES:
NB.22

$38.40


Estimated to ship onMay 19, 2025


material

polystyrene

manufacturer/tradename

Nalgene®

color

green

Looking for similar products? Visit Product Comparison Guide

General description

Colored disks fit into the recessed tops of Nalgene cryogenic vials. The system allows color-coding of vials for quick visual identification. Cut out top facilitates handling of vials with forceps. Flat surface permits writing.

Legal Information

Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service