Skip to Content

C0806

CM-Cellulose

fibers

Synonym(s):

Carboxymethyl cellulose

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
47101511
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

fibers

InChI

1S/C6H12O6.C2H4O2/c7-1-3(9)5(11)6(12)4(10)2-8;1-2(3)4/h1,3-6,8-12H,2H2;1H3,(H,3,4)

InChI key

VJHCJDRQFCCTHL-UHFFFAOYSA-N

Application

CM Cellulose products are used in protein chromatography, ion exchange chromatography and cation exchange media. CM Cellulose has been used in studies to inform the suppressed proliferation of hepatoma and leukemic cells and the inhibition of the human immunodeficiency virus type 1 (HIV-1).


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library