All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C10H7ClOS
CAS Number:
Molecular Weight:
210.68
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
SMILES string
ClCC(=O)c1csc2ccccc12
InChI
1S/C10H7ClOS/c11-5-9(12)8-6-13-10-4-2-1-3-7(8)10/h1-4,6H,5H2
InChI key
OTWORIPHJRKXBR-UHFFFAOYSA-N
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service