Skip to Content
Merck
All Photos(1)

Key Documents

C0749

Sigma-Aldrich

CAMK2γ, active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

CAMKG, MGC26678

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
51111800
NACRES:
NA.32
Pricing and availability is not currently available.

recombinant

expressed in baculovirus infected Sf9 cells

Quality Level

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

211-285 nmol/min·mg

mol wt

~60 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... CAMK2G(818)

Biochem/physiol Actions

CAMK2γ is a member of the CAMKII family which are ubiquitous serine/threonine protein kinases that have been implicated in diverse effects of hormones and neurotransmitters. CAMK2γ has six alternatively spliced variants that encode six different isoforms. Some of these variants have been identified in human tumors. Transgenic mice expressing a partially calcium-independent mutant form of CAMK2γ showed 1.5- to 2-fold increase in the thymus of these mice, at least in part due to an increase in the life span of double-positive thymocytes. There was an increase in the number of T cells in the secondary lymphoid organs that had acquired an antigen-dependent memory phenotype.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.25 mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J D Bui et al.
Cell, 100(4), 457-467 (2000-02-29)
In order to study the role of calcium/calmodulin kinase II (CaMKII) in T cells, we generated transgenic mice expressing CaMKIIgammaB* (T287D), a partially calcium-independent mutant of CaMKIIgammaB. In these mice, the size of the thymus was increased 1.5- to 2-fold
R M Tombes et al.
Biochimica et biophysica acta, 1355(3), 281-292 (1997-03-01)
CaMK-II (the (type II) multifunctional Ca2+/CaM-dependent protein kinase) has been implicated in diverse neuronal and non-neuronal functions, including cell growth control. CaMKII expression was evaluated in a variety of human tumor cell lines using RT-PCR (reverse transcriptase coupled polymerase chain

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service