Skip to Content
Merck
All Photos(1)

Key Documents

C0370

Sigma-Aldrich

CAMKK1, active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

CAMKKA, DFKZp761 M0423, MG C34095

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

recombinant

expressed in baculovirus infected Sf9 cells

Quality Level

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

9-12 nmol/min·mg

mol wt

~94 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... CAMKK1(84254)

Biochem/physiol Actions

CAMKK1 or CAMKKalpha is a Ca2+/calmodulin-dependent protein kinase that activates CAM-kinases I and IV via phosphorylation of their Thr(177) and Thr(196) residues, respectively. Recent studies have shown that the activity of CAMKK1 is decreased upon phosphorylation by cAMP-dependent protein kinase (PKA). The CAMKKalpha has been identified in intact cells as AMPKKs, predicting a significant role for this kinase in regulating AMPK activity in vivo. It has been shown that 2-deoxyglucose- and ionomycin-stimulated AMPK activity is substantially reduced in HeLa cells transfected with small interfering RNAs specific for CAMKKalpha.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.2 5mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rebecca L Hurley et al.
The Journal of biological chemistry, 280(32), 29060-29066 (2005-06-28)
The AMP-activated protein kinase (AMPK) is an important regulator of cellular metabolism in response to metabolic stress and to other regulatory signals. AMPK activity is absolutely dependent upon phosphorylation of AMPKalphaThr-172 in its activation loop by one or more AMPK
S Okuno et al.
Journal of biochemistry, 130(4), 503-513 (2001-09-28)
Ca(2+)/calmodulin-dependent protein kinases (CaM-kinases) I and IV are activated upon phosphorylation of their Thr(177) and Thr(196), respectively, by the upstream Ca(2+)/calmodulin-dependent protein kinases CaM-kinase kinase alpha and beta, and deactivated upon dephosphorylation by protein phosphatases such as CaM-kinase phosphatase. Recent

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service