BMSIPS205020E
Benchmark OS-20L Overhead Stirrer
AC/DC input 220 V, Schuko plug
Synonym(s):
overhead electric stirrer, digital overhead stirrer
All Photos(1)
About This Item
Recommended Products
packaging
pack of 1 ea
manufacturer/tradename
Benchmark Scientific IPS2050-20-E
parameter
30-2200 rpm speed
General description
Benchmark′s Overhead Stirrers feature high torque and advanced digital microprocessor control for smooth mixing of low and medium viscosity samples. Speed and run time are accurately set and monitored on the LCD screen. They are supplied with a 4 arm stainless steel propeller, additional steel and PTFE accessories are available. A feed-through chuck holds accessory shafts firmly in place and is easily tightened and loosened without tools. The overhead stirrers are protected from damage by sensors that shut down operation in the case of high viscosity or obstruction, while a splash cover protects the connection mechanism from potential corrosion. Unlike most mixers, the Benchmark series is also supplied with a support stand and mounting hardware. The stirrer easily and securely attaches to the stand for compatibility with a variety of vessels, up to 24 inches high.
Features and Benefits
- LCD control panel
- Includes complete stand assembly
- Compact and quiet
- Digital accuracy and convenience