Documents
BAH378620152
Replacement Parts for Scienceware® Hand Held Colony Counter
Red Felt Tip Pen
About This Item
UNSPSC Code:
41104000
NACRES:
NB.22
manufacturer/tradename
Bel-Art H37862-0152
General description
  • Works on plastic or glass
  • Is oil and water resistant
  • Is removable with ethyl alcohol
Legal Information
Scienceware is a registered trademark of SP Scienceware, a division of SP