Skip to Content
Merck
All Photos(2)

Key Documents

BAH136800070

Flowmi® Cell Strainers

porosity 70 μm, for 1000 uL Pipette Tips, pack of 50 ea

Synonym(s):

compact laboratory sample filters, facs sample filtration, small sample filter, small sample pipette tip filter

Sign Into View Organizational & Contract Pricing

Select a Size

50 EA
$138.00

$138.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
50 EA
$138.00

About This Item

UNSPSC Code:
41104932
NACRES:
NB.01

$138.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

material

white

sterility

sterile

packaging

pack of 50 ea
pkg of (supplied in a tray)

manufacturer/tradename

Bel-Art H13680-0070

pore size

70 μm porosity

Looking for similar products? Visit Product Comparison Guide

General description

Scienceware® Flowmi Cell Strainers provide fast and efficient filtering of small volume samples (up to 1000μl) prior to FLOW or Analysis. Patent pending.

Features and Benefits

  • Preserves volume when filtering small sample volumes
  • Decreases the potential for clogging of FLOW or FACS
  • instruments by effectively filtering cellular debris
  • Use with your favorite pipette tips; Flowmi fits most 1000μl tips including Fisherbrand Sure-One, VWR brand, Axygen, Nichiryo and Eppendorf and more
  • Compact tray holds 50 Flowmi Cell Strainers ready for press and fit attachment to your tip; features a sliding cover for easy, one-handed use
  • Packaged in a re-sealable bag to maintain sterility when used in laminar flow hoods

Other Notes

Recommended for use with samples having a maximum concentration of 4MM cells/ml.

Legal Information

Flowmi is a registered trademark of SP Scienceware, Division of SP
Scienceware is a registered trademark of SP Scienceware, a division of SP

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eduardo Javier López Soto et al.
eLife, 9 (2020-03-28)
Cell-specific alternative splicing modulates myriad cell functions and is disrupted in disease. The mechanisms governing alternative splicing are known for relatively few genes and typically focus on RNA splicing factors. In sensory neurons, cell-specific alternative splicing of the presynaptic CaV

Articles

Use of MULTI-seq lipid-modified oligos, protocol, and troubleshooting guide for PCR Assays and Sequencing applications.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service