BAB617003700
H-B® Durac® Timer
Timing capacity : 23 Hour:59 Minute, 4 channel with White Board
Synonym(s):
laboratory timer, multichannel timer, timer alarm
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
feature
4 channel with White Board
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art B61700-3700
display W × H
43.2 mm × 53.3 mm
Looking for similar products? Visit Product Comparison Guide
General description
This four channel timer has a built in white board that helps keep track of time and other critical information
Features and Benefits
Audible alarm for each channel
Built in white board and dry erase marker with eraser tip (included) helps keep track of crucial information
Programs: hours/minutes; Displays: hours/minutes/seconds
Switchable timing capacity: 23 hours: 59 minutes / 59 minutes
Count direction: up/down; displays elapsed time after countdown
Last time set memory
Key press confirmation feature gives an audible indication of data entry
Lay flat on a bench, use the flip-open stand for a vertical view, or use the magnetic back for metal surface attachment
Requires two AAA batteries (included)
Built in white board and dry erase marker with eraser tip (included) helps keep track of crucial information
Programs: hours/minutes; Displays: hours/minutes/seconds
Switchable timing capacity: 23 hours: 59 minutes / 59 minutes
Count direction: up/down; displays elapsed time after countdown
Last time set memory
Key press confirmation feature gives an audible indication of data entry
Lay flat on a bench, use the flip-open stand for a vertical view, or use the magnetic back for metal surface attachment
Requires two AAA batteries (included)
Legal Information
Durac is a registered trademark of SP Scienceware, a division of SP
H-B is a registered trademark of SP Scienceware, a division of SP
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service