Skip to Content
Merck
All Photos(1)

Key Documents

B9684

Sigma-Aldrich

Anti-Brn3a antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Brn3A Antibody, Brn3A Antibody - Anti-Brn3a antibody produced in rabbit, Anti-FLJ13449, Anti-OCT-T1, Anti-POU4F1, Anti-RDC1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 42.9 kDa

species reactivity

human

species reactivity (predicted by homology)

mouse

technique(s)

immunocytochemistry: 1:200-1:1000
immunohistochemistry (frozen sections): 1:200-1:1000
western blot: 1:200-1:1000

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... POU4F1(5457)
mouse ... Pou4f1(18996)
rat ... Pou4f1(114503)

General description

Probable transcription factor which may play a role in the regulation of specific gene expression within a subset of neuronal lineages. May play a role in determining or maintaining the identities of a small subset of visual system neurons.

Immunogen

POU4F1 / BRN3A antibody was raised against synthetic peptide (aa 141-157, AGGGGAHDGPGGGGGPG) corresponding to human Brn3a (BSA conjugated). Percent identity by BLAST analysis: Human (100%); Mouse (94%); Drosophila (82%).

Epitope
aa141-157

Physical form

Supplied in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brenna Krieger et al.
PloS one, 12(7), e0180091-e0180091 (2017-07-29)
The retina communicates with the brain using ≥30 parallel channels, each carried by axons of distinct types of retinal ganglion cells. In every mammalian retina one finds so-called "alpha" ganglion cells (αRGCs), identified by their large cell bodies, stout axons
Peter Stacey et al.
SLAS discovery : advancing life sciences R & D, 23(6), 585-596 (2018-03-17)
Screening against a disease-relevant phenotype to identify compounds that change the outcome of biological pathways, rather than just the activity of specific targets, offers an alternative approach to find modulators of disease characteristics. However, in pain research, use of in
RAGE and its ligand amyloid beta promote retinal ganglion cell loss following ischemia-reperfusion injury.
Seyed Hosseini Fin, et al.
Frontiers in Cellular Neuroscience, 17, 1156084-1156084 (2023)
Fuxin Shi et al.
The European journal of neuroscience, 26(11), 3016-3023 (2007-11-17)
In mammals, hair cells and auditory neurons lack the capacity to regenerate, and damage to either cell type can result in hearing loss. Replacement cells for regeneration could potentially be made by directed differentiation of human embryonic stem (hES) cells.
Hans Jürgen Wester et al.
Theranostics, 5(6), 618-630 (2015-04-01)
Chemokine ligand-receptor interactions play a pivotal role in cell attraction and cellular trafficking, both in normal tissue homeostasis and in disease. In cancer, chemokine receptor-4 (CXCR4) expression is an adverse prognostic factor. Early clinical studies suggest that targeting CXCR4 with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service