Skip to Content
Merck

Skip To

B9561

Monoclonal Anti-Brain-derived Neurotrophic Factor antibody produced in mouse

clone 35909, purified immunoglobulin, lyophilized powder

Synonym(s):

Anti-BDNF

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
51111800
NACRES:
NA.43
MDL number:
Conjugate:
unconjugated
Clone:
35909, monoclonal
Application:
FACS
Citations:
6
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

35909, monoclonal

form

lyophilized powder

species reactivity

human

technique(s)

flow cytometry: 25 μg/mL

isotype

IgG1

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BDNF(627)

General description

Brain derived neurotrophic factor (BDNF) belongs to a family of secreted proteins called neurotrophins that includes, nerve growth factor (NGF), neurotrophin-3 (NT3) and NT4/5. BDNF is expressed in the developing and the mature brain. The influence of BDNF is mediated by its binding to TrkB receptor and sometimes the receptor p75NTR. The downstream pathways promoted by BDNF include PI3K, MAPK and JAK/STAT. The most important role of BDNF is the regulation of synaptic transmission and plasticity, protein synthesis and phosphorylation of local proteins. The secretion of BDNF is highly regulated at both transcriptional and translational levels since it is involved in the proper function of neuronal circuit and development of cognitive functions. The deficits in BDNF functions are implicated in a number of neurodegenerative disorders such as Huntington′s disease, schizophrenia and dementia
Monoclonal Anti-Brain-derived Neurotrophic Factor detects human BDNF.

Immunogen

purified, Sf21-derived recombinant human brain-derived neurotrophic factor.

Application

Monoclonal Anti-Brain-derived Neurotrophic Factor may be used to detect human BDNF by flow cytometry using a working antibody concentration of 25 μg/ml.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline with 5% trehalose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

or

Try our Product Selector Tool to narrow your options


Storage Class

13 - Non Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters