Skip to Content
Merck
All Photos(1)

Key Documents

B9384

Sigma-Aldrich

2-(2-Bromoethyl)-1,3-dioxane

Synonym(s):

3-Bromopropionaldehyde trimethylene acetal

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11BrO2
CAS Number:
Molecular Weight:
195.05
Beilstein:
1421628
EC Number:
MDL number:
UNSPSC Code:
12352100

refractive index

n20/D 1.481 (lit.)

bp

67-70 °C/2.8 mmHg (lit.)

density

1.431 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

BrCCC1OCCCO1

InChI

1S/C6H11BrO2/c7-3-2-6-8-4-1-5-9-6/h6H,1-5H2

InChI key

WMDHQEHPOVOEOG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

206.6 °F - closed cup

Flash Point(C)

97 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service