Skip to Content
Merck
All Photos(1)

Documents

B9270

Sigma-Aldrich

Blue Trisacryl® M

aqueous ethanol suspension, 40-80 μm

Synonym(s):

Blue Trisacryl®

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
13111000
NACRES:
NA.56

form

aqueous ethanol suspension

matrix

beaded macroporous copolymer of N-tris(hydroxymethyl)methylacrylamide

matrix spacer

6 atoms

particle size

40-80 μm

capacity

≥10 mg/mL binding capacity (human serum albumin)

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Blue Trisacryl is used in affinity chromatography and protein chromatography. Blue Trisacryl has been used to purify transferrin from human plasma and to study immune response.

Physical form

Suspension in 1 M NaCl containing 20% ethanol

Legal Information

Trisacryl is a registered trademark of Pall Corporation

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

176.0 °F

Flash Point(C)

80 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Use of blue-Trisacryl M for purifying transferrin from human plasma].
B Buatois et al.
Annales pharmaceutiques francaises, 45(4), 321-328 (1987-01-01)
Younes Achouri et al.
The Biochemical journal, 381(Pt 1), 35-42 (2004-04-09)
Extracts of frozen rat liver were found to catalyse the formation of 3H2O from DL-2-hydroxy[2-3H]glutarate. Three peaks of enzyme activities were observed on separation by chromatography on DEAE-Sepharose. The first and second peaks corresponded to an enzyme acting on L-2-hydroxyglutarate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service