Skip to Content
Merck
All Photos(2)

Key Documents

B9180

Sigma-Aldrich

BW A868C

≥98% (HPLC)

Synonym(s):

3-[(2-Cyclohexyl-2-hydroxyethyl)amino]-2,5-dioxo-1-(phenylmethyl)- 4-imidazolidineheptanoic acid

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$825.00

$825.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$825.00

About This Item

Empirical Formula (Hill Notation):
C25H37N3O5
CAS Number:
Molecular Weight:
459.58
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$825.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk Order

Assay

≥98% (HPLC)

form

film

color

colorless

solubility

DMSO: 5 mg/mL, clear

storage temp.

−20°C

SMILES string

OC(CNN1C(CCCCCCC(O)=O)C(=O)N(Cc2ccccc2)C1=O)C3CCCCC3

InChI

1S/C25H37N3O5/c29-22(20-13-7-4-8-14-20)17-26-28-21(15-9-1-2-10-16-23(30)31)24(32)27(25(28)33)18-19-11-5-3-6-12-19/h3,5-6,11-12,20-22,26,29H,1-2,4,7-10,13-18H2,(H,30,31)

InChI key

YZJVWSKJHGEIBL-UHFFFAOYSA-N

Biochem/physiol Actions

BW A868C is a potent, selective DP prostanoid receptor antagonist.

Features and Benefits

This compound is featured on the Prostanoid Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Oliveira et al.
British journal of pharmacology, 129(3), 509-514 (2000-03-11)
1. Cumulative concentration-effect curves for the selective prostanoid TP receptor agonist U46619 and six isoprostanes were constructed in the human isolated umbilical artery. 2. All compounds except 8-iso-PGF3 alpha produced concentration-dependent contractions. The contractile response to the isoprostanes increased with
Hui Zhao et al.
Peptides, 29(3), 345-349 (2008-01-09)
Met-Arg-Trp (MRW) has been isolated as an inhibitor for angiotensin I-converting enzyme (ACE) from a pepsin-pancreatin digest of spinach ribulose bisphosphate carboxylase/oxygenase (Rubisco) (IC(50)=0.6 microM). It has been reported that hypotensive activity of ACE-inhibitory peptides derived from food proteins are
T Minami et al.
The Journal of pharmacology and experimental therapeutics, 278(3), 1146-1152 (1996-09-01)
We previously reported that intrathecal administration of prostaglandin (PG) D2 and PGE2 to conscious mice induced hyperalgesia (assessed by a hot-plate test) and that intrathecal administration of PGE2 and PGF2 alpha induced allodynia, a state of discomfort and pain evoked
Md Ashequr Rahman et al.
European journal of pharmacology, 567(3), 245-251 (2007-06-05)
The present study was undertaken to investigate the involvement of chemical mediators in nasal allergic responses using histidine decarboxylase knockout (HDC-KO) mice. An allergic rhinitis model was developed in HDC-KO and wild-type mice by the intraperitoneal injection of ovalbumin, aluminum
N A Sharif et al.
British journal of pharmacology, 131(6), 1025-1038 (2000-11-18)
1. A potent and highly selective DP prostanoid receptor antagonist radioligand, [(3)H]-cyclohexyl-N-BWA868C (3-benzyl-5-(6-carboxyhexyl)-1-(2-cyclohexyl-2-hydroxyethyl-amino) hydantoin, ([(3)H]-BWA868C)), has been generated for receptor binding and autoradiographic studies. 2. Specific [(3)H]-BWA868C binding to human platelet membranes achieved equilibrium within 60 min at 23 degrees

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service