Skip to Content
Merck

Skip To

B7937

BSc3094 monohydrobromide

≥98% (HPLC)

Promotion Available

Synonym(s):

2-[4-(4-Nitrophenyl)-2-thiazolyl]hydrazide-1H-benzimidazole-6-carboxylic acid monohydrobromide

Sign In to View Organizational & Contract Pricing.

Spend ARS 530,000 — Save 5%; Spend ARS 907,000 — Save 10%; Spend ARS 1,500,000 — Save 22%*

Sign in to check eligibility. Shop Products

Select a Size

Change View
Size/SKUAvailabilityPrice
5 mg
Check Cart for Availability
$253.00

About This Item

Empirical Formula (Hill Notation):
C17H12N6O3S·HBr
CAS Number:
Molecular Weight:
461.29
NACRES:
NA.25
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:
Assay:
≥98% (HPLC)
Form:
solid
Storage condition:
protect from light

$253.00

PRICE WITHOUT NATIONAL TAXES

Save Up To 22%


Check Cart for Availability

​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

solid

storage condition

protect from light

solubility

DMSO: 22 mg/mL

storage temp.

2-8°C

SMILES string

Br.[O-][N+](=O)c1ccc(cc1)-c2csc(NNC(=O)c3ccc4[nH]cnc4c3)n2

InChI

1S/C17H12N6O3S.BrH/c24-16(11-3-6-13-14(7-11)19-9-18-13)21-22-17-20-15(8-27-17)10-1-4-12(5-2-10)23(25)26;/h1-9H,(H,18,19)(H,20,22)(H,21,24);1H

InChI key

HVNYYJJWBWMGCO-UHFFFAOYSA-N

Application

BSc3094 is used in tau protein amyloidogenicity/tauopathy research to study the processes of tau aggregation and cross-linking in neurodegenerative disease development.

Biochem/physiol Actions

BSc3094 is a potent inhibitor of tau aggregation and dissolves preformed tau paired helical filaments. BSc3094 interacts closely with the tau protein at the edge involving protons I-IV, while the second attachment site seems to be at the nitro group. In N2A cells (model system for tau aggregation), BSc3094 exhibited low toxicity.
BSc3094 is a potent inhibitor of tau aggregation.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
S5696SML2922SML0077
description

≥98% (HPLC)

description

≥98% (HPLC), solid

description

≥98% (HPLC)

description

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

form

solid

form

solid

form

powder

form

powder

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

solubility

DMSO: 22 mg/mL

solubility

DMSO: ≥20 mg/mL

solubility

DMSO: 2 mg/mL, clear

solubility

DMSO: ≥12 mg/mL

storage condition

protect from light

storage condition

protect from light

storage condition

-

storage condition

-


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

dust mask type N95 (US), Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters