Skip to Content
Merck
All Photos(1)

Key Documents

B7381

Sigma-Aldrich

1,4-Butanediyl diglycidyl ether

~70%

Synonym(s):

Araldite® RD-2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18O4
CAS Number:
Molecular Weight:
202.25
Beilstein:
115238
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

~70%

bp

155-160 °C/11 mmHg

density

1.05 g/mL (lit.)

SMILES string

C(CCOCC1CO1)COCC2CO2

InChI

1S/C10H18O4/c1(3-11-5-9-7-13-9)2-4-12-6-10-8-14-10/h9-10H,1-8H2

InChI key

SHKUUQIDMUMQQK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Araldite is a registered trademark of Huntsman Advanced Materials Inc.

replaced by

Product No.
Description
Pricing

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P E Morgan et al.
Journal of molecular recognition : JMR, 9(5-6), 394-400 (1996-09-01)
The preparation, characterisation and testing of stable non-porous coated perfluorocarbon supports functionalised with the metal chelate, iminodiacetic acid (IDA) is described. Polyvinyl alcohol (PVA), a neutral hydrophilic polymer was esterified with perfluorooctanoyl chloride and anchored to the surface of solid
The new face of fillers: why evidence and experience both count.
Hema Sundaram
Journal of drugs in dermatology : JDD, 10(9), 964-964 (2011-11-05)
P B van Wachem et al.
Journal of biomedical materials research, 55(3), 415-423 (2001-03-20)
Calcification limits the long-term durability of xenograft glutaraldehyde (GA)-crosslinked heart valves. Previously, a study in rats showed that epoxy-crosslinked heart valves reduced lymphocyte reactions to the same extent as the GA-crosslinked control and induced a similar foreign-body response and calcification
David Stocks et al.
Journal of drugs in dermatology : JDD, 10(9), 974-980 (2011-11-05)
Hyaluronic acid (HA) gels are commonly injected into the skin to lift rhytides and to improve facial appearance. The different processes used in their manufacture and formulation yield products with unique physical characteristics that play an important role in predicting
M M Diogo et al.
Bioseparation, 10(4-5), 211-220 (2002-09-18)
This work presents studies on the interactions of supercoiled plasmid DNA and Escherichia coli genomic DNA (gDNA) and RNA, with an hydrophobic interaction chromatography (HIC) gel, obtained by derivatisation of Sepharose CL-6B with 1,4-butanediol diglycidyl ether. Nucleic acids purified from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service