Skip to Content
Merck
All Photos(1)

Key Documents

B6930

Sigma-Aldrich

Nalgene® dropper bottle with screw closure

capacity 8 mL, natural bottle, assorted colors cap

Synonym(s):

bottle dropper, dropper bottles

Sign Into View Organizational & Contract Pricing

Select a Size

25 EA
$91.90

$91.90

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
25 EA
$91.90

About This Item

UNSPSC Code:
41121800
NACRES:
NB.17

$91.90

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

material

low-density polyethylene
natural bottle
round bottle

Joint

Joints threaded neck (15)

packaging

pack of 25 ea

manufacturer/tradename

Nalgene® 2752-9025

capacity

8 mL

color

assorted colors cap

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

  • Nalgene® dropper bottles with screw closure is an alternative to various pipetting devices.
  • The tip ensures accurate dispensing of 40μL drops (based on water; viscosity affects drop size), dispensed one drop at a time.
  • Chemically resistant to most reagents.
  • LDPE clear transparent bottles for easy sample visualization.
  • Leak-proof closure - prevents sample loss.
  • Control dispensing tip ensures a secure fit.
  • Suitable for use in biotech, diagnostic and pharmaceutical applications.

Legal Information

Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service