Skip to Content
Merck

Skip To

B6535

Nalgene® bottles, style 2114

capacity 250 mL

Synonym(s):

HDPE bottles, nalgene bottle, nalgene bottles, Nalgene 2114-0008, laboratory container, vessel

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
12 ea
Check Cart for Availability
$136.00

About This Item

NACRES:
NB.17
UNSPSC Code:
41121800

$136.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

high-density polyethylene bottle, polypropylene cap, polypropylene screw closure, square translucent bottle

joint

Joints threaded neck (43)

description

Not autoclavable

feature

with cap

packaging

pack of 12 ea

manufacturer/tradename

Nalgene® 2114-0008

bottle L × H × W

61 mm × 116 mm × 61 mm

capacity

250 mL

General description

Easily fill square wide-mouth bottles. HDPE exhibits excellent chemical resistance to most acids, bases and alcohols, making these bottles suitable for a wide variety of uses. Good for freezer storage to -100°C (-148°F). This product is ACT Labeled by My Green Lab to enable data-based sustainable product purchasing decisions.

Features and Benefits

  • Wide mouth is easy to fill.
  • Bottle design saves space in freezer.
  • HDPE exhibits excellent chemical resistance to most acids, bases and alcohols, making these bottles suitable for a wide variety of uses.
  • Linerless caps provide leakproof† protection without the use of a liner that can wrinkle, cause leaks, or contaminate your reagents.
  • Leakproof† when used with Nalgene closures.
  • Bottle and Carton are readily recyclable in most communities.
  • Manufactured from non-toxic materials.
  • Participates in My Green Lab′s ACT Label program for Accountability, Consistency and Transparency (ACT) around evaluating product sustainability

Legal Information

Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
B6410B6660B6785
material

high-density polyethylene bottle, polypropylene screw closure, polypropylene cap, square translucent bottle

material

high-density polyethylene bottle, polypropylene cap, polypropylene screw closure, square translucent bottle

material

high-density polyethylene bottle, polypropylene cap, polypropylene screw closure, square translucent bottle

material

high-density polyethylene bottle, polypropylene cap, polypropylene screw closure, square translucent bottle

manufacturer/tradename

Nalgene® 2114-0008

manufacturer/tradename

Nalgene® 2114-0006

manufacturer/tradename

Nalgene® 2114-0016

manufacturer/tradename

Nalgene® 2114-0032

feature

with cap

feature

with cap

feature

with cap

feature

with cap

capacity

250 mL

capacity

175 mL

capacity

500 mL

capacity

1,000 mL

joint

Joints threaded neck (43)

joint

Joints threaded neck (38-415)

joint

Joints threaded neck (53)

joint

Joints threaded neck (63)

packaging

pack of 12 ea

packaging

pack of 12 ea

packaging

pack of 12 ea

packaging

pack of 6 ea


Still not finding the right product?

Explore all of our products under Nalgene® bottles, style 2114




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library