Skip to Content
Merck

Skip To

B6060

BVT.948

≥98% (HPLC), solid

Synonym(s):

4-Hydroxy-3,3-dimethyl-2H-benzo[g]indole-2,5(3H)-dione

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 mg
Check Cart for Availability
$263.00
$223.55

About This Item

Empirical Formula (Hill Notation):
C14H11NO3
CAS Number:
Molecular Weight:
241.24
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:
Assay:
≥98% (HPLC)
Form:
solid
Storage condition:
desiccated, protect from light, under inert gas

$223.55

List Price$263.00Save 15%
PRICE WITHOUT NATIONAL TAXES
Web-Only Promotion

Check Cart for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

solid

storage condition

desiccated, protect from light, under inert gas

color

red

solubility

DMSO: 18 mg/mL, H2O: insoluble

storage temp.

−20°C

SMILES string

CC1(C)C(=O)N=C2c3ccccc3C(=O)C(O)=C12

InChI

1S/C14H11NO3/c1-14(2)9-10(15-13(14)18)7-5-3-4-6-8(7)11(16)12(9)17/h3-6,17H,1-2H3

InChI key

BAQXWJQSUXZVIP-UHFFFAOYSA-N

Biochem/physiol Actions

BVT.948 is a non-competitive inhibitor of protein tyrosine phosphatase (PTP). Enhances insulin signaling in cells and inhibits several cytochrome P450 isoforms in vitro.

Features and Benefits

BVT.948 is a non-competitive inhibitor of protein tyrosine phosphatase (PTP). Enhances insulin signaling in cells and inhibits several cytochrome P450 isoforms in vitro. Works through a different mechanism then the other PTP inhibitors. BVT.948 was able to enhance insulin signaling in cells, although it did not increase tyrosine phosphorylation globally. Furthermore, the compound was active in vivo, enhancing insulin tolerance tests in ob/ob mice, therefore apparently enhancing insulin sensitivity. BVT.948 was able to inhibit several other PTPs tested and also was efficient at inhibiting several cytochrome P450 isoforms in vitro.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
L4170S8442D3943
description

≥98% (HPLC), solid

description

≥98% (HPLC)

description

≥98% (HPLC)

description

≥98% (HPLC), solid

form

solid

form

solid

form

powder

form

solid

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

2-8°C

storage condition

desiccated, under inert gas, protect from light

storage condition

desiccated, protect from light

storage condition

-

storage condition

-

solubility

DMSO: 18 mg/mL, H2O: insoluble

solubility

DMSO: 16 mg/mL

solubility

DMSO: 20 mg/mL, clear (warmed), H2O: insoluble

solubility

DMSO: >20 mg/mL, H2O: insoluble


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters