Skip to Content
Merck
All Photos(2)

Key Documents

B5438

Sigma-Aldrich

Balsalazide disodium salt hydrate

Synonym(s):

5-[4-(2-Carboxyethylcarbamoyl)phenylazo]salicylic acid disodium salt hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H13N3Na2O6 · xH2O
CAS Number:
Molecular Weight:
401.28 (anhydrous basis)
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.25

Assay

≥99% (HPLC)

form

powder

storage temp.

2-8°C

SMILES string

O.[Na+].[Na+].Oc1ccc(cc1C([O-])=O)\N=N\c2ccc(cc2)C(=O)NCCC([O-])=O

InChI

1S/C17H15N3O6.2Na.H2O/c21-14-6-5-12(9-13(14)17(25)26)20-19-11-3-1-10(2-4-11)16(24)18-8-7-15(22)23;;;/h1-6,9,21H,7-8H2,(H,18,24)(H,22,23)(H,25,26);;;1H2/q;2*+1;/p-2/b20-19+;;;

InChI key

ZZZXQZHSFPVNCR-HVARVCSCSA-L

Biochem/physiol Actions

Balsalazide has antioxidative and anti-inflammatory properties. It inhibits superoxide production and was found to scavenge hypochlorite radicals. Balsalazide was shown to inhibit lipid peroxidation in rat intestinal mucosa. It was also shown to inhibit azoxymethane-induced colon carcinogenesis. In vivo, basalazide is cleaved through azoreduction to produce 4-aminobenzoyl-β-alanine, an inert carrier moiety, and 5-aminosalicylic acid (5-ASA).

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service