Skip to Content
Merck
All Photos(2)

Key Documents

B4932

Sigma-Aldrich

Butyl-γ-cyclodextrin

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

form

solid

storage temp.

2-8°C

SMILES string

CCCCOC[C@H]1O[C@@H]2O[C@H]3[C@H](O)[C@@H](O)[C@H](O[C@@H]3CO)O[C@H]4[C@H](O)[C@@H](O)[C@H](O[C@@H]4CO)O[C@H]5[C@H](O)[C@@H](O)[C@H](O[C@@H]5CO)O[C@H]6[C@H](O)[C@@H](O)[C@H](O[C@@H]6CO)O[C@H]7[C@H](O)[C@@H](O)[C@H](O[C@@H]7CO)O[C@H]8[C@H](O)[C@@H](O)[C@H](O[C@@H]8CO)O[C@H]9[C@H](O)[C@@H](O)[C@H](O[C@@H]9CO)O[C@H]1[C@H](O)[C@H]2O

InChI

1S/C52H88O40/c1-2-3-4-76-12-20-44-28(67)36(75)52(84-20)91-43-19(11-59)82-50(34(73)26(43)65)89-41-17(9-57)80-48(32(71)24(41)63)87-39-15(7-55)78-46(30(69)22(39)61)85-37-13(5-53)77-45(29(68)21(37)60)86-38-14(6-54)79-47(31(70)23(38)62)88-40-16(8-56)81-49(33(72)25(40)64)90-42-18(10-58)83-51(92-44)35(74)27(42)66/h13-75H,2-12H2,1H3/t13-,14-,15-,16-,17-,18-,19-,20-,21-,22-,23-,24-,25-,26-,27-,28-,29-,30-,31-,32-,33-,34-,35-,36-,37-,38-,39-,40-,41-,42-,43-,44-,45-,46-,47-,48-,49-,50-,51-,52-/m1/s1

InChI key

GKKAPJLYIDRKJM-CEIOSFLZSA-N

Other Notes

To gain a comprehensive understanding of our extensive range of Oligosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Hu et al.
Methods and findings in experimental and clinical pharmacology, 31(7), 449-456 (2009-11-13)
Owing to their bioadaptability and multifunctional characteristics, cyclodextrins have been shown to be useful in drug delivery systems. This article presents the unique structure and properties and the current application of natural and chemically modified cyclodextrins in transdermal drug delivery
Kaneto Uekama
Chemical & pharmaceutical bulletin, 52(8), 900-915 (2004-08-12)
The pharmaceutically useful cyclodextrins (CyDs) are classified into hydrophilic, hydrophobic, and ionic derivatives. Because of the multi-functional characteristics and bioadaptability, these CyDs are capable of alleviating the undesirable properties of drug molecules through the formation of inclusion complexes or the
Birna S Gudmundsdottir et al.
Journal of ocular pharmacology and therapeutics : the official journal of the Association for Ocular Pharmacology and Therapeutics, 30(1), 35-41 (2013-11-12)
To test a new drug delivery platform with dorzolamide γ-cyclodextrin (γCD) nanoparticle eye drops for intraocular pressure (IOP) control and safety and compare with Trusopt.(®) Self-aggregating γCD nanoparticle eye drops containing 3% dorzolamide were given once a day (QD) and
Feng Zhao et al.
Biomaterials, 35(3), 1050-1062 (2013-11-06)
For developing a multifunctional bioreducible targeted and synergistic co-delivery system for anticancer drug paclitaxel (PTX) and p53 gene for potential cancer therapy, supramolecular self-assembled inclusion complex was prepared from PTX and star-shaped cationic polymer containing γ-cyclodextrin (γ-CD) and multiple oligoethylenimine
Prosenjit Bhattacharya et al.
Journal of fluorescence, 24(3), 719-725 (2013-12-18)
This paper reports the double confinement of 4,4-diaminodiphenyl sulfone (Dapsone) inside γ-cyclodextrin (CD) in presence of surfactants (cationic, anionic and nonionic) using steady-state and time-resolved fluorescence spectroscopy. Interpretation of fluorescence spectra, fluorescence anisotropy and time resolved fluorescence decay of the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service