Skip to Content
Merck
All Photos(1)

Key Documents

B4813

Sigma-Aldrich

BGP-15

≥98% (HPLC)

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$219.00
25 MG
$878.00

$219.00


Estimated to ship onMay 23, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$219.00
25 MG
$878.00

About This Item

Empirical Formula (Hill Notation):
C14H22N4O2· 2HCl
CAS Number:
Molecular Weight:
351.27
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$219.00


Estimated to ship onMay 23, 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

solid

solubility

deionized water: 28 mg/mL

storage temp.

−20°C

SMILES string

Cl.Cl.N\C(=N/OCC(O)CN1CCCCC1)c2cccnc2

InChI

1S/C14H22N4O2.2ClH/c15-14(12-5-4-6-16-9-12)17-20-11-13(19)10-18-7-2-1-3-8-18;;/h4-6,9,13,19H,1-3,7-8,10-11H2,(H2,15,17);2*1H

InChI key

ISGGVCWFTPTHIX-UHFFFAOYSA-N

General description

BGP-15 is a hydroxylamine derivative, which has cytoprotective and neuroprotective properties. It reduces phospho- c-Jun N-terminal kinase (pJNK) and p38 stress signalling. BGP-15 also stimulates non-specific serine/threonine protein kinase (AKT) and insulin-like growth factor 1 receptor (IGFR1) protective signalling.[1] BGP-15 functions as an insulin sensitizer in an olanzapine-induced metabolic disorder. It protects cells against cell death and enhances the expression of heat shock protein (HSP).[2] BGP-15 might be used to treat heart failure (HF) and atrial fibrillation (AF).[3]

Application

BGP-15 has been used to study its influence on proteasomal degradation of missense dystrophins.[4]

Biochem/physiol Actions

BGP-15 is PARP inhibitor, HSP72 activator.
BGP-15 is PARP inhibitor, HSP72 activator.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Disease-proportional proteasomal degradation of missense dystrophins
Talsness DM, et al.
Proceedings of the National Academy of Sciences of the USA, 112(40), 12414-12419 (2015)
BGP-15 prevents the death of neurons in a mouse model of familial dysautonomia
Ohlen SB, et al.
Proceedings of the National Academy of Sciences of the USA, 114(19), 5035-5040 (2017)
BGP-15 protects against oxidative stress-or lipopolysaccharide-induced mitochondrial destabilization and reduces mitochondrial production of reactive oxygen species
Sumegi K, et al.
PLoS ONE, 12(1), e0169372-e0169372 (2017)
Urszula Brykczynska et al.
Cell reports, 30(5), 1627-1643 (2020-02-06)
The innate immune system safeguards the organism from both pathogenic and environmental stressors. Also, physiologic levels of nutrients affect organismal and intra-cellular metabolism and challenge the immune system. In the long term, over-nutrition leads to low-grade systemic inflammation. Here, we
Matthew S Elitt et al.
Nature, 585(7825), 397-403 (2020-07-02)
Mutations in PLP1, the gene that encodes proteolipid protein (PLP), result in failure of myelination and neurological dysfunction in the X-chromosome-linked leukodystrophy Pelizaeus-Merzbacher disease (PMD)1,2. Most PLP1 mutations, including point mutations and supernumerary copy variants, lead to severe and fatal disease.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service