Skip to Content
Merck
All Photos(1)

Key Documents

B4437

Sigma-Aldrich

p38β, active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

MAPK11, PRKM11, SAPK2, SAPK2B, p38-2, p38b

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf9 cells

Quality Level

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

105-141 nmol/min·mg

mol wt

~71 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... MAPK11(5600)

Biochem/physiol Actions

p38β is a member of the p38 MAP kinase family and is activated by both proinflammatory cytokines and environmental stress. The p38β is activated through its phosphorylation by MAP kinase kinases (MKKs), preferably by MKK6. Transcription factor ATF2/CREB2 has been shown to be a substrate of this kinase.Alternatively spliced transcript variants encoding the same protein have been observed.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.2 5mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Jiang et al.
The Journal of biological chemistry, 271(30), 17920-17926 (1996-07-26)
Mitogen-activated protein (MAP) kinase cascades represent one of the major signal systems used by eukaryotic cells to transduce extracellular signals into cellular responses. Four MAP kinase subgroups have been identified in humans: ERK, JNK (SAPK), ERK5 (BMK), and p38. Here
B Stein et al.
The Journal of biological chemistry, 272(31), 19509-19517 (1997-08-01)
Mitogen-activated protein (MAP) kinases are involved in many cellular processes. Here we describe the cloning and characterization of a new MAP kinase, p38-2. p38-2 belongs to the p38 subfamily of MAP kinases and shares with it the TGY phosphorylation motif.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service